Assay Details
Target Gene Details
Entrez Gene ID: | 67841 |
Gene Name: | autophagy related 3 |
Gene Aliases: |
2610016C12Rik, APG3, Apg3l, Atg3l, PC3-96 |
Location: |
Chr.16:45158829-45188538 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atg3 | NM_026402.3 | NP_080678.1 | ||
| AB079386.1 | BAC57452.1 | |||
| AK005266.1 | BAB23918.1 | |||
| AK011409.1 | BAB27600.1 | |||
| AK078877.1 | BAC37437.1 | |||
| AK150914.1 | BAE29952.1 | |||
| AK159231.1 | BAE34917.1 | |||
| AK209780.1 | ||||
| AK213960.1 | ||||
| AK220135.1 | ||||
| BC010809.1 | AAH10809.1 |
More Information
Set Membership: |
|

Genomic Map