Assay Details
Target Gene Details
Entrez Gene ID: | 13356 |
Gene Name: | DiGeorge syndrome critical region gene 2 |
Gene Aliases: |
9930034O06Rik, DGS-C, Dgsc, Idd, Lan, Sez12, mKIAA0163 |
Location: |
Chr.16:17840355-17894098 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dgcr2 | NM_001109750.1 | NP_001103220.1 | ||
| NM_010048.3 | NP_034178.2 | |||
| AK042786.1 | ||||
| AK088131.1 | BAC40164.1 | |||
| AK129070.1 | BAC97880.1 | |||
| AK141594.1 | BAE24753.1 | |||
| AK145495.1 | BAE26471.1 | |||
| AK153026.1 | ||||
| AK161359.1 | ||||
| AK163872.1 | BAE37523.1 | |||
| AK163903.1 | ||||
| AK193194.1 | ||||
| AK215211.1 | ||||
| BC060636.1 | AAH60636.1 | |||
| BC062978.1 | AAH62978.1 | |||
| D78641.1 | BAA11460.1 | |||
| X95480.1 | CAA64749.1 |

Genomic Map