Assay Details
Target Gene Details
Entrez Gene ID: | 13356 |
Gene Name: | DiGeorge syndrome critical region gene 2 |
Gene Aliases: |
9930034O06Rik, DGS-C, Dgsc, Idd, Lan, Sez12, mKIAA0163 |
Location: |
Chr.16:17840355-17894098 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dgcr2 | NM_001109750.1 | 6 | 965 | NP_001103220.1 |
| NM_010048.3 | 6 | 974 | NP_034178.2 | |
| AK042786.1 | 6 | 896 | ||
| AK088131.1 | 6 | 895 | BAC40164.1 | |
| AK129070.1 | BAC97880.1 | |||
| AK133880.1 | 3 | 263 | BAE21909.1 | |
| AK141594.1 | 6 | 940 | BAE24753.1 | |
| AK145495.1 | 6 | 891 | BAE26471.1 | |
| AK153026.1 | 6 | 762 | ||
| AK161359.1 | 6 | 690 | ||
| AK163872.1 | 6 | 927 | BAE37523.1 | |
| AK163903.1 | 6 | 902 | ||
| AK190458.1 | ||||
| AK194718.1 | ||||
| AK201465.1 | ||||
| AK202091.1 | ||||
| BC060636.1 | 6 | 915 | AAH60636.1 | |
| BC062978.1 | 6 | 869 | AAH62978.1 | |
| D78641.1 | 6 | 899 | BAA11460.1 | |
| X95480.1 | 6 | 840 | CAA64749.1 |

Genomic Map