Assay Details
Target Gene Details
Entrez Gene ID: | 13544 |
Gene Name: | dishevelled 3, dsh homolog (Drosophila) |
Gene Aliases: |
- |
Location: |
Chr.16:20516938-20532548 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dvl3 | NM_007889.2 | NP_031915.2 | ||
| AK052151.1 | ||||
| AK220155.1 | 2 | 4281 | BAD90341.1 | |
| U41285.1 | AAB01761.1 |
More Information
Set Membership: |
|

Genomic Map