Assay Details
Target Gene Details
Entrez Gene ID: | 223723 |
Gene Name: | tubulin tyrosine ligase-like family, member 12 |
Gene Aliases: |
BC055368, D430005B17 |
Location: |
Chr.15:83575094-83595157 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttll12 | NM_183017.2 | NP_898838.2 | ||
| AK084879.1 | ||||
| AK134345.1 | ||||
| AK143496.1 | ||||
| AK150116.1 | BAE29319.1 | |||
| AM690756.1 | ||||
| BC055368.1 | AAH55368.1 |

Genomic Map