Assay Details
Target Gene Details
Entrez Gene ID: | 380993 |
Gene Name: | zinc finger and AT hook domain containing |
Gene Aliases: |
Gm922, Zfat1, Zfp406 |
Location: |
Chr.15:68083714-68259265 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfat | NM_001145888.1 | 10 | 2910 | NP_001139360.1 |
| NM_198644.2 | 10 | 2889 | NP_941046.1 | |
| BC053930.1 | 10 | 2880 | AAH53930.1 | |
| EU221277.1 | 10 | 2740 |
More Information
Set Membership: |
|

Genomic Map