Assay Details
Target Gene Details
Entrez Gene ID: | 57781 |
Gene Name: | CD200 receptor 1 |
Gene Aliases: |
CD200R, Mox2r, OX2R |
Location: |
Chr.16:44765736-44794977 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd200r1 | NM_021325.3 | NP_067300.1 | ||
| AK054236.1 | ||||
| AK089173.1 | BAC40777.1 | |||
| AK089415.1 | BAC40872.1 | |||
| AK154325.1 | BAE32516.1 | |||
| AK154722.1 | BAE32786.1 | |||
| AK170735.1 | ||||
| AK171130.1 | BAE42266.1 | |||
| AK171590.1 | BAE42546.1 | |||
| BC052682.1 | AAH52682.1 |
More Information
Set Membership: |
|

Genomic Map