Assay Details
Target Gene Details
Entrez Gene ID: | 271375 |
Gene Name: | Cd200 receptor 2 |
Gene Aliases: |
AY230198, Cd200r1l |
Location: |
Chr.16:44867097-44915840 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd200r2 | NM_206535.1 | NP_996258.1 | ||
| AY230198.1 | AAO84052.1 |
More Information
Set Membership: |
|

Genomic Map