Assay Details
Target Gene Details
Entrez Gene ID: | 223723 |
Gene Name: | tubulin tyrosine ligase-like family, member 12 |
Gene Aliases: |
BC055368, D430005B17 |
Location: |
Chr.15:83575094-83595157 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttll12 | NM_183017.2 | 9 | 1318 | NP_898838.2 |
| AK084879.1 | 9 | 1285 | ||
| AK134345.1 | 9 | 1285 | ||
| AK143496.1 | 8 | 1150 | ||
| AK150116.1 | 9 | 1284 | BAE29319.1 | |
| AM690756.1 | 9 | 1271 | ||
| BC055368.1 | 9 | 1283 | AAH55368.1 |

Genomic Map