Assay Details
Target Gene Details
Entrez Gene ID: | 319953 |
Gene Name: | tubulin tyrosine ligase-like 1 |
Gene Aliases: |
6330444E16Rik, AV014541 |
Location: |
Chr.15:83483769-83518917 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttll1 | NM_178869.4 | NP_849200.2 | ||
| AK031917.1 | BAC27603.1 | |||
| AK047046.1 | BAC32947.1 | |||
| AK084005.1 | BAC39092.1 | |||
| AK140472.1 | BAE24404.1 | |||
| AK168786.1 | BAE40620.1 | |||
| AK169150.1 | BAE40929.1 | |||
| BC010510.1 | AAH10510.1 |
More Information
Set Membership: |
|

Genomic Map