Assay Details
Target Gene Details
Entrez Gene ID: | 319953 |
Gene Name: | tubulin tyrosine ligase-like 1 |
Gene Aliases: |
6330444E16Rik, AV014541 |
Location: |
Chr.15:83483769-83518917 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttll1 | NM_178869.4 | 8 | 1082 | NP_849200.2 |
| AK031917.1 | 8 | 1053 | BAC27603.1 | |
| AK047046.1 | 8 | 1067 | BAC32947.1 | |
| AK084005.1 | 8 | 1050 | BAC39092.1 | |
| AK140472.1 | 8 | 1050 | BAE24404.1 | |
| AK168786.1 | 8 | 1053 | BAE40620.1 | |
| AK169150.1 | 8 | 1069 | BAE40929.1 | |
| AK209983.1 | ||||
| BC010510.1 | 8 | 1031 | AAH10510.1 |
More Information
Set Membership: |
|

Genomic Map