Assay Details
Target Gene Details
Entrez Gene ID: | 239591 |
Gene Name: | tubulin tyrosine ligase-like family, member 8 |
Gene Aliases: |
1700019P01Rik, 4933401B17 |
Location: |
Chr.15:88889778-88955641 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttll8 | NM_172818.3 | NP_766406.2 | ||
| AK029745.1 | BAC26595.1 | |||
| AK030151.1 | BAC26811.1 | |||
| AM690752.1 | ||||
| BC116293.1 | ||||
| BC116294.1 |

Genomic Map