Assay Details
Target Gene Details
Entrez Gene ID: | 239591 |
Gene Name: | tubulin tyrosine ligase-like family, member 8 |
Gene Aliases: |
1700019P01Rik, 4933401B17 |
Location: |
Chr.15:88889778-88955641 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttll8 | NM_172818.3 | 8 | 1364 | NP_766406.2 |
| AK029745.1 | 8 | 1360 | BAC26595.1 | |
| AK030151.1 | 8 | 1357 | BAC26811.1 | |
| AM690752.1 | 8 | 1021 | ||
| BC116293.1 | 8 | 1005 | ||
| BC116294.1 | 8 | 1000 |

Genomic Map