Assay Details
Target Gene Details
Entrez Gene ID: | 22143 |
Gene Name: | tubulin, alpha 1B |
Gene Aliases: |
Tuba2 |
Location: |
Chr.15:98931427-98934560 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tuba1b | AK075955.1 | BAC36080.1 | ||
| AK137885.1 | BAE23512.1 | |||
| AK150128.1 | BAE29327.1 | |||
| AK150179.1 | BAE29362.1 | |||
| AK150968.1 | BAE29999.1 | |||
| AK151199.1 | BAE30196.1 | |||
| AK151809.1 | BAE30708.1 | |||
| AK152298.1 | BAE31107.1 | |||
| AK153064.1 | BAE31690.1 | |||
| AK153254.1 | BAE31845.1 | |||
| AK158958.1 | 1 | 116 | BAE34741.1 | |
| AK164428.1 | BAE37783.1 | |||
| AK168770.1 | BAE40606.1 | |||
| AK169075.1 | BAE40860.1 | |||
| AK169092.1 | BAE40875.1 | |||
| AK169130.1 | BAE40909.1 | |||
| AK169665.1 | BAE41287.1 | |||
| BC002219.1 | AAH02219.1 | |||
| BC008117.1 | AAH08117.1 | |||
| BC083120.1 | AAH83120.1 | |||
| BC098321.1 | AAH98321.1 | |||
| BC108337.1 | AAI08338.1 | |||
| M13446.1 | AAA40500.1 |

Genomic Map