Assay Details
Target Gene Details
Entrez Gene ID: | 68744 |
Gene Name: | zinc finger protein 740 |
Gene Aliases: |
1110034O07Rik, AW548228, Znf740 |
Location: |
Chr.15:102203645-102215610 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp740 | NM_001289690.1 | NP_001276619.1 | ||
| NM_001289691.1 | NP_001276620.1 | |||
| NM_001289694.1 | NP_001276623.1 | |||
| NM_001289695.1 | NP_001276624.1 | |||
| AK144707.1 | BAE26025.1 | |||
| AK170207.1 | ||||
| AK180920.1 | ||||
| AK183663.1 | ||||
| AK190632.1 | ||||
| BC030410.1 | AAH30410.1 | |||
| BC066011.1 | AAH66011.1 | |||
| CA319591.1 |
More Information
Set Membership: |
|

Genomic Map