Assay Details
Target Gene Details
Entrez Gene ID: | 106389 |
Gene Name: | ELL associated factor 2 |
Gene Aliases: |
AW048865, FESTA-L, FESTA-S, Festa, Traits, U19 |
Location: |
Chr.16:36792884-36874826 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eaf2 | NM_134111.2 | NP_598872.1 | ||
| AK076871.1 | ||||
| AK132343.1 | ||||
| BC056626.1 | AAH56626.1 |

Genomic Map