Assay Details
Target Gene Details
Entrez Gene ID: | 106389 |
Gene Name: | ELL associated factor 2 |
Gene Aliases: |
AW048865, FESTA-L, FESTA-S, Festa, Traits, U19 |
Location: |
Chr.16:36792884-36874826 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eaf2 | NM_001113401.1 | NP_001106872.1 | ||
| NM_001113405.1 | NP_001106876.1 | |||
| NM_134111.2 | NP_598872.1 | |||
| AB081298.1 | BAC77525.1 | |||
| AK076871.1 | ||||
| AY034479.1 | AAK59701.1 | |||
| AY049021.1 | AAL12224.1 | |||
| BC004721.1 | AAH04721.1 | |||
| BC056626.1 | AAH56626.1 |

Genomic Map