Assay Details
Target Gene Details
Entrez Gene ID: | 22763 |
Gene Name: | zinc finger RNA binding protein |
Gene Aliases: |
C920030H05Rik |
Location: |
Chr.15:12117793-12185449 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfr | NM_011767.2 | NP_035897.2 | ||
| AF071059.1 | AAC25762.1 | |||
| AK034963.1 | BAC28897.1 | |||
| AK158938.1 | BAE34733.1 | |||
| BC141139.1 |
More Information
Set Membership: |
|

Genomic Map