Assay Details
Target Gene Details
Entrez Gene ID: | 210544 |
Gene Name: | TBC1 domain family, member 31 |
Gene Aliases: |
4B3, D330013L20Rik, Gm85, Wdr67 |
Location: |
Chr.15:57911684-57970068 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tbc1d31 | NM_001081396.2 | NP_001074865.2 | ||
| NM_001167679.1 | NP_001161151.1 | |||
| AB185844.1 | BAD83881.1 | |||
| AK084542.1 | BAC39214.1 | |||
| AK141805.1 | BAE24839.1 | |||
| AK154264.1 | BAE32475.1 | |||
| AK161403.1 | ||||
| AK171024.1 | BAE42193.1 | |||
| BC050039.1 | AAH50039.1 | |||
| BC066830.1 | AAH66830.1 |

Genomic Map