Assay Details
Target Gene Details
Entrez Gene ID: | 56722 |
Gene Name: | LPS-induced TN factor |
Gene Aliases: |
3222402J11Rik, C85531, N4WBP3, TBX1 |
Location: |
Chr.16:10959273-10993121 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Litaf | NM_019980.2 | 2 | 149 | NP_064364.1 |
| AF171100.1 | 2 | 117 | AAF27312.1 | |
| AF220207.1 | 1 | 71 | AAG44246.1 | |
| AF230522.1 | 1 | 99 | AAO49168.1 | |
| AK018578.1 | 2 | 132 | BAB31289.1 | |
| AK076162.1 | 2 | 151 | BAC36228.1 | |
| AK078708.1 | 2 | 149 | ||
| AK150476.1 | ||||
| AK151053.1 | 2 | 149 | BAE30070.1 | |
| AK159533.1 | 2 | 117 | BAE35161.1 | |
| BC018559.1 | 2 | 129 | AAH18559.1 |
More Information
Set Membership: |
|

Genomic Map