Assay Details
Target Gene Details
Entrez Gene ID: | 30060 |
Gene Name: | antigen p97 (melanoma associated) identified by monoclonal antibodies 133.2 and 96.5 |
Gene Aliases: |
CD228, MTf |
Location: |
Chr.16:31878501-31899020 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mfi2 | NM_013900.2 | NP_038928.1 | ||
| AB024336.1 | BAA86655.1 | |||
| AK036444.1 | BAC29432.1 | |||
| BC040347.1 | AAH40347.1 |
More Information
Set Membership: |
|

Genomic Map