Assay Details
Target Gene Details
Entrez Gene ID: | 67824 |
Gene Name: | NmrA-like family domain containing 1 |
Gene Aliases: |
1110025F24Rik, AI256624 |
Location: |
Chr.16:4711317-4719356 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nmral1 | NM_001290761.1 | NP_001277690.1 | ||
| NM_001290762.1 | NP_001277691.1 | |||
| NM_001290763.1 | NP_001277692.1 | |||
| NM_026393.2 | NP_080669.1 | |||
| AK012340.1 | BAB28172.1 | |||
| AK078389.1 | BAC37249.1 | |||
| AK195249.1 | ||||
| BC030039.1 | AAH30039.1 |

Genomic Map