Assay Details
Target Gene Details
Entrez Gene ID: | 71338 |
Gene Name: | transformation related protein 63 regulated |
Gene Aliases: |
5430420C16Rik, Tprg1 |
Location: |
Chr.16:25286817-25423588 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tprg | NM_175165.3 | NP_780374.1 | ||
| AK017323.1 | ||||
| AK036796.1 | BAC29579.1 |

Genomic Map