Assay Details
Target Gene Details
Entrez Gene ID: | 71338 |
Gene Name: | transformation related protein 63 regulated |
Gene Aliases: |
5430420C16Rik, Tprg1 |
Location: |
Chr.16:25286817-25423588 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tprg | NM_175165.3 | 5 | 742 | NP_780374.1 |
| AK036796.1 | 5 | 745 | BAC29579.1 |

Genomic Map