Assay Details
Target Gene Details
Entrez Gene ID: | 20440 |
Gene Name: | beta galactoside alpha 2,6 sialyltransferase 1 |
Gene Aliases: |
AW742324, Siat1, St6Gal-I, St6gal, St6galI |
Location: |
Chr.16:23224728-23360350 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| St6gal1 | NM_001252505.1 | 2 | 260 | NP_001239434.1 |
| NM_145933.4 | 2 | 591 | NP_666045.1 | |
| AF153681.1 | 1 | 98 | ||
| AK084124.1 | 2 | 426 | BAC39120.1 | |
| BC092222.1 | 2 | 569 | AAH92222.1 | |
| BC096026.1 | 2 | 239 | AAH96026.1 | |
| BY254258.1 | 2 | 261 | ||
| CD349601.1 | 2 | 229 | ||
| CF898349.1 |
More Information
Set Membership: |
|

Genomic Map