Assay Details
Target Gene Details
Entrez Gene ID: | 26425 |
Gene Name: | nucleotide binding protein 1 |
Gene Aliases: |
- |
Location: |
Chr.16:10411878-10424428 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nubp1 | NM_011955.2 | NP_036085.1 | ||
| AF114170.1 | AAF01786.1 | |||
| AK020141.1 | BAB32007.1 | |||
| AK157212.1 | BAE34002.1 | |||
| AK178734.1 | ||||
| BC055436.1 | AAH55436.1 | |||
| BU614991.1 | ||||
| CX210766.1 |
Target Gene Details
Entrez Gene ID: | 383103 |
Gene Name: | trans-golgi network vesicle protein 23A |
Gene Aliases: |
Fam18a |
Location: |
Chr.16:10420557-10447388 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tvp23a | NM_001013778.1 | NP_001013800.1 | ||
| BC068110.1 | AAH68110.1 |

Genomic Map