Assay Details
Target Gene Details
Entrez Gene ID: | 26425 |
Gene Name: | nucleotide binding protein 1 |
Gene Aliases: |
- |
Location: |
Chr.16:10411878-10424428 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nubp1 | NM_011955.2 | NP_036085.1 | ||
| AF114170.1 | AAF01786.1 | |||
| AK020876.1 | BAB32236.1 | |||
| AK157212.1 | BAE34002.1 | |||
| BC055436.1 | AAH55436.1 | |||
| CJ183745.1 | ||||
| CX210766.1 |

Genomic Map