Assay Details
Target Gene Details
Entrez Gene ID: | 65079 |
Gene Name: | reticulon 4 receptor |
Gene Aliases: |
NOGOR, NgR, NgR1 |
Location: |
Chr.16:18127706-18152408 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rtn4r | NM_022982.2 | NP_075358.1 | ||
| AF283462.1 | AAG53611.1 | |||
| BC052317.1 | AAH52317.2 | |||
| BC058381.1 | AAH58381.1 |

Genomic Map