Assay Details
Target Gene Details
Entrez Gene ID: | 20440 |
Gene Name: | beta galactoside alpha 2,6 sialyltransferase 1 |
Gene Aliases: |
AW742324, Siat1, St6Gal-I, St6gal, St6galI |
Location: |
Chr.16:23224728-23360350 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| St6gal1 | NM_001252505.1 | NP_001239434.1 | ||
| NM_001252506.1 | NP_001239435.1 | |||
| NM_145933.4 | NP_666045.1 | |||
| AB027197.1 | BAB00636.1 | |||
| AK084124.1 | BAC39120.1 | |||
| BC027833.1 | AAH27833.1 | |||
| BC092222.1 | AAH92222.1 | |||
| BC096026.1 | AAH96026.1 | |||
| CF741963.1 | ||||
| CN458528.1 | ||||
| D16106.1 | BAA03680.1 |
More Information
Set Membership: |
|

Genomic Map