Assay Details
Target Gene Details
Entrez Gene ID: | 14852 |
Gene Name: | G1 to S phase transition 1 |
Gene Aliases: |
AI314175, AI326449, AV307676, C79774, G1st, Gst-1 |
Location: |
Chr.16:11216240-11254325 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gspt1 | NM_001130008.1 | NP_001123480.1 | ||
| NM_146066.2 | NP_666178.2 | |||
| AB003502.1 | BAA32526.1 | |||
| AK030760.1 | ||||
| AK032059.1 | BAC27675.1 | |||
| AK075996.1 | BAC36107.1 | |||
| AK145785.1 | BAE26649.1 | |||
| AK148603.1 | ||||
| AK166626.1 | ||||
| BC028325.1 | AAH28325.1 | |||
| BC031640.1 | AAH31640.1 |
More Information
Set Membership: |
|

Genomic Map