Assay Details
Target Gene Details
Entrez Gene ID: | 320007 |
Gene Name: | SID1 transmembrane family, member 1 |
Gene Aliases: |
AW045928, B830021E24Rik |
Location: |
Chr.16:44240177-44333328 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 23 - Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sidt1 | NM_001159419.1 | NP_001152891.1 | ||
| NM_198034.3 | NP_932151.2 | |||
| AK034344.1 | BAC28682.1 | |||
| AK046836.1 | ||||
| AK171920.1 | ||||
| AY562213.1 | AAS87378.1 | |||
| AY562214.1 | AAS87379.1 | |||
| BC025888.1 | AAH25888.1 | |||
| BC079584.1 | AAH79584.1 |

Genomic Map