Assay Details
Target Gene Details
Entrez Gene ID: | 66067 |
Gene Name: | GTP-binding protein 8 (putative) |
Gene Aliases: |
0610037H22Rik |
Location: |
Chr.16:44738879-44746377 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gtpbp8 | NM_001159329.1 | 5 | 554 | NP_001152801.1 |
| NM_025332.3 | 5 | 755 | NP_079608.1 | |
| AK002771.1 | 5 | 740 | ||
| AK010979.1 | 5 | 752 | BAB27301.1 | |
| AK084919.1 | 1 | 1569 | ||
| AK147840.1 | 5 | 739 | BAE28175.1 | |
| BC107216.1 | 5 | 752 | AAI07217.1 | |
| BC107217.1 | 5 | 551 | AAI07218.1 |

Genomic Map