Assay Details
Target Gene Details
Entrez Gene ID: | 16509 |
Gene Name: | potassium voltage-gated channel, Isk-related subfamily, member 1 |
Gene Aliases: |
Isk, MinK, nmf190 |
Location: |
Chr.16:92345995-92359468 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcne1 | NM_008424.3 | NP_032450.1 | ||
| AK008938.1 | BAB25979.1 | |||
| AK028907.1 | BAC26189.1 | |||
| BC094276.1 | AAH94276.1 |
More Information
Set Membership: |
|

Genomic Map