Assay Details
Target Gene Details
Entrez Gene ID: | 15233 |
Gene Name: | homogentisate 1, 2-dioxygenase |
Gene Aliases: |
Hgo, aku |
Location: |
Chr.16:37580153-37632026 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hgd | NM_013547.3 | 13 | 1274 | NP_038575.2 |
| NR_027778.1 | 12 | 1185 | ||
| AK002584.1 | 12 | 1147 | ||
| BC024369.1 | 12 | 1088 | ||
| BC037628.1 | 13 | 1188 | AAH37628.2 | |
| BC044758.2 | 12 | 1074 | ||
| BC055029.1 | 13 | 1155 | AAH55029.1 |
More Information
Set Membership: |
|

Genomic Map