Assay Details
Target Gene Details
Entrez Gene ID: | 17450 |
Gene Name: | microrchidia 1 |
Gene Aliases: |
Morc |
Location: |
Chr.16:48431237-48630905 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Morc1 | NM_010816.1 | NP_034946.1 | ||
| AF084945.1 | AAD43003.1 | |||
| BC030893.1 | AAH30893.1 |

Genomic Map