Assay Details
Target Gene Details
Entrez Gene ID: | 106407 |
Gene Name: | solute carrier family 51, alpha subunit |
Gene Aliases: |
AV001382, AW261577, D630035O19Rik, OSTalpha, Osta |
Location: |
Chr.16:32475578-32487879 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc51a | NM_145932.3 | NP_666044.1 | ||
| AK052725.1 | BAC35116.1 | |||
| BC024441.1 | AAH24441.1 | |||
| BC025912.1 | AAH25912.1 | |||
| BC031178.1 | AAH31178.1 |
More Information
Set Membership: |
|

Genomic Map