Assay Details
Target Gene Details
Entrez Gene ID: | 277203 |
Gene Name: | transmembrane 4 L six family member 19 |
Gene Aliases: |
EG277203 |
Location: |
Chr.16:32400012-32408579 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tm4sf19 | NM_001160402.1 | NP_001153874.1 | ||
| BC147727.1 | ||||
| BC147729.1 |

Genomic Map