Assay Details
Target Gene Details
Entrez Gene ID: | 72388 |
Gene Name: | receptor-interacting serine-threonine kinase 4 |
Gene Aliases: |
2310069J12Rik, AI552420, ANKK2, Ankrd3, DIk, PKK, RIP4 |
Location: |
Chr.16:97741933-97763755 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ripk4 | NM_023663.6 | NP_076152.2 | ||
| AF302127.3 | AAG30871.2 | |||
| AK077233.1 | ||||
| AK145203.1 | BAE26294.1 | |||
| BC057871.1 | AAH57871.1 |
More Information
Set Membership: |
|

Genomic Map