Assay Details
Target Gene Details
Entrez Gene ID: | 66863 |
Gene Name: | leucine-zipper-like transcriptional regulator, 1 |
Gene Aliases: |
1200003E21Rik, AI591627, AW550890 |
Location: |
Chr.16:17508541-17526333 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lztr1 | NM_025808.3 | 9 | 988 | NP_080084.2 |
| AK004561.2 | 9 | 988 | BAB23373.2 | |
| AK005037.1 | 9 | 987 | BAB23764.2 | |
| AK083411.1 | 9 | 992 | BAC38905.1 | |
| AK132817.1 | 9 | 987 | BAE21377.1 | |
| BC034400.1 | 9 | 985 | AAH34400.1 |
More Information
Set Membership: |
|

Genomic Map