Assay Details
Target Gene Details
Entrez Gene ID: | 20357 |
Gene Name: | sema domain, seven thrombospondin repeats (type 1 and type 1-like), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 5B |
Gene Aliases: |
AI893641, SemG, Semag, mKIAA1445 |
Location: |
Chr.16:35541154-35664261 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sema5b | NM_013661.2 | NP_038689.2 | ||
| AK140524.1 | 5 | 1146 | ||
| BC052397.1 | AAH52397.1 | |||
| X97818.1 | CAA66398.1 |
More Information
Set Membership: |
|

Genomic Map