Assay Details
Target Gene Details
Entrez Gene ID: | 57738 |
Gene Name: | solute carrier family 15 (H+/peptide transporter), member 2 |
Gene Aliases: |
8430408C16Rik, C78862, Pept2 |
Location: |
Chr.16:36750164-36785157 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc15a2 | NM_001145899.1 | NP_001139371.1 | ||
| NM_021301.3 | NP_067276.2 | |||
| AF111811.1 | AAF42470.1 | |||
| AK018393.1 | BAB31192.1 | |||
| AK128939.1 | ||||
| BC018335.1 | AAH18335.1 | |||
| BC051199.1 | AAH51199.1 | |||
| BC138627.1 | ||||
| BC138628.1 | ||||
| CA469599.1 | ||||
| JF495121.1 | ||||
| JF495122.1 |
More Information
Set Membership: |
|

Genomic Map