Assay Details
Target Gene Details
Entrez Gene ID: | 66664 |
Gene Name: | transmembrane protein 41a |
Gene Aliases: |
2900010K02Rik, 5730578N08Rik |
Location: |
Chr.16:21934327-21947552 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem41a | NM_025693.4 | NP_079969.1 | ||
| NR_037773.1 | ||||
| AK007685.1 | BAB25189.1 | |||
| AK013518.1 | BAB28894.1 | |||
| AK160292.1 | ||||
| BB853849.1 | ||||
| BB873279.1 | ||||
| BC019770.1 | AAH19770.1 |

Genomic Map