Assay Details
Target Gene Details
Entrez Gene ID: | 66664 |
Gene Name: | transmembrane protein 41a |
Gene Aliases: |
2900010K02Rik, 5730578N08Rik |
Location: |
Chr.16:21934327-21947552 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem41a | NM_025693.4 | 3 | 416 | NP_079969.1 |
| NR_037773.1 | 2 | 262 | ||
| AK007685.1 | 3 | 411 | BAB25189.1 | |
| AK013518.1 | 2 | 241 | BAB28894.1 | |
| AK160292.1 | 3 | 337 | ||
| BB853849.1 | 3 | 416 | ||
| BC019770.1 | 3 | 371 | AAH19770.1 |

Genomic Map