Assay Details
Target Gene Details
Entrez Gene ID: | 71751 |
Gene Name: | mitogen-activated protein kinase kinase kinase 13 |
Gene Aliases: |
C130026N12Rik, LZK |
Location: |
Chr.16:21825171-21933439 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Map3k13 | NM_172821.3 | NP_766409.2 | ||
| AK047986.1 | ||||
| AK051378.1 | BAC34618.1 | |||
| DQ480427.1 |
More Information
Set Membership: |
|

Genomic Map