Assay Details
Target Gene Details
Entrez Gene ID: | 68092 |
Gene Name: | nuclear cap binding protein subunit 2 |
Gene Aliases: |
20kDa, 5930413E18Rik, AA536802, AI843301, C79367 |
Location: |
Chr.16:31948546-31958472 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ncbp2 | NM_026554.4 | NP_080830.1 | ||
| BY035864.1 | 1 | 48 |
More Information
Set Membership: |
|

Genomic Map