Assay Details
Target Gene Details
Entrez Gene ID: | 207227 |
Gene Name: | syntaxin binding protein 5-like |
Gene Aliases: |
9430094L02, A830015P08Rik, LLGL4, T2dm1, t2md1, tomosyn-2 |
Location: |
Chr.16:37107307-37384988 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stxbp5l | NM_001114611.1 | NP_001108083.1 | ||
| NM_001114612.1 | NP_001108084.1 | |||
| NM_001114613.1 | NP_001108085.1 | |||
| NM_172440.3 | NP_766028.2 | |||
| AK035159.1 | BAC28964.1 | |||
| AY542324.1 | AAT68175.1 | |||
| AY542325.1 | AAT68176.1 | |||
| AY542326.1 | AAT68177.1 | |||
| AY542327.1 | AAT68178.1 | |||
| BC137883.1 | ||||
| BC145007.1 | ||||
| BC145008.1 | ||||
| BC145009.1 |
More Information
Set Membership: |
|

Genomic Map