Assay Details
Target Gene Details
Entrez Gene ID: | 207227 |
Gene Name: | syntaxin binding protein 5-like |
Gene Aliases: |
9430094L02, A830015P08Rik, LLGL4, T2dm1, t2md1, tomosyn-2 |
Location: |
Chr.16:37107307-37384988 on Build GRCm38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stxbp5l | NM_001114611.1 | 24 | 3154 | NP_001108083.1 |
| NM_001114612.1 | 25 | 3325 | NP_001108084.1 | |
| NM_001114613.1 | 25 | 3226 | NP_001108085.1 | |
| NM_172440.3 | 26 | 3397 | NP_766028.2 | |
| AY542324.1 | 25 | 3248 | AAT68175.1 | |
| AY542325.1 | 23 | 3005 | AAT68176.1 | |
| AY542326.1 | 24 | 3176 | AAT68177.1 | |
| AY542327.1 | 24 | 3077 | AAT68178.1 | |
| BU609516.1 | 3 | 468 |
More Information
Set Membership: |
|

Genomic Map