Assay Details
Target Gene Details
Entrez Gene ID: | 67203 |
Gene Name: | nuclear distribution gene E homolog 1 (A nidulans) |
Gene Aliases: |
2810027M15Rik, AU042936, AW822251, Nude, mNudE |
Location: |
Chr.16:14163275-14192928 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nde1 | NM_001114085.1 | NP_001107557.1 | ||
| NM_001285503.1 | NP_001272432.1 | |||
| NM_001285504.1 | NP_001272433.1 | |||
| NM_023317.2 | NP_075806.2 | |||
| AF290473.1 | AAG10062.1 | |||
| AF322073.1 | AAK08067.1 | |||
| AK012830.1 | BAB28499.1 | |||
| AK012861.1 | ||||
| AK037289.1 | BAE20509.1 | |||
| AK146982.1 | BAE27585.1 | |||
| AK150826.1 | BAE29888.1 | |||
| AK166858.1 | ||||
| AK169743.1 | BAE41342.1 | |||
| BC023267.1 | AAH23267.1 |

Genomic Map