Assay Details
Target Gene Details
Entrez Gene ID: | 68194 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1 beta subcomplex 4 |
Gene Aliases: |
0610006N12Rik, 1300010H20Rik |
Location: |
Chr.16:37647602-37654368 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufb4 | NM_026610.1 | NP_080886.1 | ||
| AK004958.1 | 1 | 180 | BAB23699.1 | |
| AK009807.1 | BAB26515.1 | |||
| AK018707.1 | BAB31358.1 | |||
| BC056229.1 | AAH56229.1 | |||
| BC096421.1 | AAH96421.1 |
More Information
Set Membership: |
|

Genomic Map