Assay Details
Target Gene Details
Entrez Gene ID: | 73916 |
Gene Name: | intraflagellar transport 57 |
Gene Aliases: |
4833420A15Rik, Esrrbl1, Hippi, MHS4R2 |
Location: |
Chr.16:49699294-49765126 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ift57 | NM_028680.3 | NP_082956.2 | ||
| AF274590.1 | AAK69403.1 | |||
| AF357350.1 | ||||
| AK014731.1 | BAB29522.1 | |||
| AK040328.1 | ||||
| AK047217.1 | BAC32995.1 | |||
| BC138143.1 | ||||
| BC138144.1 | ||||
| DQ266091.1 | ABB72788.1 |
More Information
Set Membership: |
|

Genomic Map